Gene Transcripts

Gene Symbol
Vmn2r74
Entrez Gene ID
546980
Gene Name
vomeronasal 2, receptor 74
Gene Aliases
EG546980
Chromosome Location
Chr. 7: 85951867 - 85961482 on Build GRCm38
UniGene ID
Mm.469905
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105187.1 NP_001098657.1 6 2540

View Full Product Details