Gene Transcripts

Gene Symbol
Mstn
Entrez Gene ID
29152
Gene Name
myostatin
Gene Aliases
Gdf8
Chromosome Location
Chr. 9: 53310977 - 53315804 on Build Rnor_6.0
UniGene ID
Rn.44460
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019151.1 NP_062024.1 3 926
GenBank mRNA AF019624.1 - 3 926
FQ224469.1 - 1050

View Full Product Details