Gene Transcripts

Gene Symbol
Vma21
Entrez Gene ID
67048
Gene Name
VMA21 vacuolar H+-ATPase homolog (S. cerevisiae)
Gene Aliases
2610030H06Rik, AI840175
Chromosome Location
Chr. X: 71812752 - 71824706 on Build GRCm38
UniGene ID
Mm.371816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081356.3 NP_001074825.1 3 2135
NM_001290780.1 NP_001277709.1 3 2262
NM_001290781.1 NP_001277710.1 4 2436
GenBank mRNA AK045401.1 - 3 2093
AK164163.1 - 3 2090

View Full Product Details