Gene Transcripts

Gene Symbol
Rabep2
Entrez Gene ID
80754
Gene Name
rabaptin, RAB GTPase binding effector protein 2
Gene Aliases
Fra, MP13
Chromosome Location
Chr. 1: 197839583 - 197855953 on Build Rnor_6.0
UniGene ID
Rn.3228
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030585.1 NP_085074.1 3, 4 461
GenBank mRNA BC089795.1 - 3, 4 447
U34932.1 - 3, 4 461

View Full Product Details