Gene Transcripts

Gene Symbol
Tnfsf10
Entrez Gene ID
246775
Gene Name
tumor necrosis factor (ligand) superfamily, member 10
Gene Aliases
Trail
Chromosome Location
Chr. 2: 113008324 - 113024535 on Build Rnor_6.0
UniGene ID
Rn.83627
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145681.1 NP_663714.1 1, 2 152
GenBank mRNA AY115578.1 - 1, 2 152
EF030546.1 - 1, 2 143

View Full Product Details