Gene Transcripts

Gene Symbol
Thumpd3
Entrez Gene ID
14911
Gene Name
THUMP domain containing 3
Gene Aliases
AW556087, Gt(ROSA)26asSor, Gtrosa26as
Chromosome Location
Chr. 6: 113046252 - 113068278 on Build GRCm38
UniGene ID
Mm.781
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008188.2 NP_032214.1 2, 3 362
GenBank mRNA AK017246.1 - 2, 3 375
AK142140.1 - 2, 3 381
BC002024.1 - 2, 3 364 3591186
BC012688.1 - 1, 2 310 3987438
BC046800.1 - 2, 3 362 6403880
U83176.1 - 2, 3 356

View Full Product Details