Gene Transcripts

Gene Symbol
Kifc2
Entrez Gene ID
16581
Gene Name
kinesin family member C2
Gene Aliases
Chromosome Location
Chr. 15: 76659837 - 76668196 on Build GRCm38
UniGene ID
Mm.378950
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010630.2 NP_034760.2 5 601
GenBank mRNA AK144304.1 - 6 760
D49545.1 - 5 677
U92949.1 - 5 591

View Full Product Details