Gene Transcripts
- Gene Symbol
- Ldha
- Entrez Gene ID
- 16828
- Gene Name
- lactate dehydrogenase A
- Gene Aliases
- Ldh1, Ldhm, l7R2
- Chromosome Location
- Chr. 7: 46845804 - 46855627 on Build GRCm38
- UniGene ID
- Mm.29324
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001136069.2 |
NP_001129541.2
|
7, 8 |
1173 |
|
NM_010699.2 |
NP_034829.1
|
7, 8 |
980 |
|
GenBank mRNA |
AK089929.1 |
- |
7, 8 |
944 |
|
AK146121.1 |
- |
7, 8 |
941 |
|
AK146980.1 |
- |
3, 4 |
355 |
|
AK149919.1 |
- |
7, 8 |
941 |
|
AK150156.1 |
- |
7, 8 |
944 |
|
AK167789.1 |
- |
7, 8 |
942 |
|
AK167944.1 |
- |
7, 8 |
942 |
|
AK168346.1 |
- |
7, 8 |
941 |
|
AK168920.1 |
- |
7, 8 |
946 |
|
AK170386.1 |
- |
7, 8 |
945 |
|
AK170708.1 |
- |
7, 8 |
942 |
|
AK171373.1 |
- |
6, 7 |
825 |
|
AK200149.1 |
- |
3, 4 |
289 |
|
AK206536.1 |
- |
1, 2 |
38 |
|
AK207234.1 |
- |
1, 2 |
38 |
|
AK207750.1 |
- |
1, 2 |
38 |
|
AK207846.1 |
- |
1, 2 |
38 |
|
AK212267.1 |
- |
1, 2 |
38 |
|
AK217640.1 |
- |
1, 2 |
38 |
|
AK219220.1 |
- |
1, 2 |
38 |
|
BC005509.1 |
- |
6, 7 |
792 |
|
BC066858.1 |
- |
6, 7 |
598 |
|
BC094019.1 |
- |
7, 8 |
897 |
4195220 |
BC094428.1 |
- |
7, 8 |
934 |
30700835 |
M17516.1 |
- |
2, 3 |
239 |
|
U13687.1 |
- |
7, 8 |
1040 |
|
- Gene Symbol
- Anxa6
- Entrez Gene ID
- 11749
- Gene Name
- annexin A6
- Gene Aliases
- AW107198, Anx6, AnxVI, Cabm, Camb
- Chromosome Location
- Chr. 11: 54978962 - 55033471 on Build GRCm38
- UniGene ID
- Mm.265347
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK203372.1 |
- |
|
278 |
|
View Full Product Details