Gene Transcripts

Gene Symbol
Tenm2
Entrez Gene ID
23964
Gene Name
teneurin transmembrane protein 2
Gene Aliases
Odz2, Odz3, Ten-2, Ten-m2
Chromosome Location
Chr. 11: 36006656 - 37236808 on Build GRCm38
UniGene ID
Mm.439889
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290702.1 NP_001277631.1 1 224
NM_011856.4 NP_035986.3 1 224
GenBank mRNA AB025411.1 - 1 261
BC141361.1 - 1 225 9055973

View Full Product Details