Gene Transcripts
- Gene Symbol
- Snrpa
- Entrez Gene ID
- 53607
- Gene Name
- small nuclear ribonucleoprotein polypeptide A
- Gene Aliases
- C430021M15Rik, Rnu1a-1, Rnu1a1, U1-A, U1A
- Chromosome Location
- Chr. 7: 27187006 - 27196271 on Build GRCm38
- UniGene ID
- Mm.386890
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001046637.1 |
NP_001040102.1
|
1, 2 |
321 |
|
NM_001285825.1 |
NP_001272754.1
|
2, 3 |
395 |
|
NM_015782.3 |
NP_056597.3
|
2, 3 |
398 |
|
GenBank mRNA |
AK013886.1 |
- |
2, 3 |
372 |
|
AK088550.1 |
- |
2, 3 |
256 |
|
BC003229.1 |
- |
2, 3 |
298 |
3483391 |
BC094006.1 |
- |
2, 3 |
258 |
4983854 |
BC096648.1 |
- |
2, 3 |
252 |
1397390 |
CA552113.1 |
- |
2, 3 |
284 |
|
CX206945.1 |
- |
2, 3 |
398 |
|
CX565356.1 |
- |
1, 2 |
291 |
|
DV650448.1 |
- |
|
14 |
|
L15447.1 |
- |
2, 3 |
194 |
|
View Full Product Details