Gene Transcripts

Gene Symbol
Nedd9
Entrez Gene ID
18003
Gene Name
neural precursor cell expressed, developmentally down-regulated gene 9
Gene Aliases
Cas-L, CasL, HEF1, MEF1, Nedd-9, p105
Chromosome Location
Chr. 13: 41309916 - 41487360 on Build GRCm38
UniGene ID
Mm.288980
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111324.2 NP_001104794.1 3 442
NM_017464.5 NP_059492.3 2 223
GenBank mRNA AF009366.1 - 3 378
AK030985.1 - 3 376
AK033729.1 - 2 224
AK046357.1 - 2 186
AK054179.1 - 2 219
AK083374.1 - 3 370

View Full Product Details