Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005923.3 | NP_005914.1 | 5 | 1211 | |
XM_011535839.2 | XP_011534141.2 | 6 | 1535 | ||
XM_017010870.1 | XP_016866359.1 | 6 | 1534 | ||
XM_017010871.1 | XP_016866360.1 | 6 | 1534 | ||
XM_017010872.1 | XP_016866361.1 | 6 | 1533 | ||
XM_017010873.1 | XP_016866362.1 | 6 | 1532 | ||
XM_017010874.1 | XP_016866363.1 | 6 | 1534 | ||
XM_017010875.1 | XP_016866364.1 | 6 | 1533 | ||
XM_017010876.1 | XP_016866365.1 | 4 | 542 | ||
XM_017010877.1 | XP_016866366.1 | 4 | 779 | ||
GenBank mRNA | AB209093.1 | - | 5 | 1315 | |
AK293953.1 | - | 5 | 471 | ||
AK310942.1 | - | 6 | 990 | ||
BC054503.2 | - | 5 | 1158 | 6143181 | |
BC088829.1 | - | 5 | 1212 | 6007002 | |
D84476.2 | - | 5 | 1117 | ||
U67156.1 | - | 5 | 1210 |