Gene Transcripts
- Gene Symbol
- KCTD15
- Entrez Gene ID
- 79047
- Gene Name
- potassium channel tetramerization domain containing 15
- Gene Aliases
- Chromosome Location
- Chr. 19: 33795540 - 33815761 on Build GRCh38
- UniGene ID
- Hs.221873
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001129994.1 |
NP_001123466.1
|
4, 5 |
518 |
|
NM_001129995.1 |
NP_001123467.1
|
4, 5 |
515 |
|
NM_024076.2 |
NP_076981.2
|
4, 5 |
518 |
|
XM_011527296.1 |
XP_011525598.1
|
4, 5 |
501 |
|
XM_011527297.2 |
XP_011525599.1
|
5, 6 |
552 |
|
XM_011527298.2 |
XP_011525600.1
|
5, 6 |
587 |
|
XM_017027283.1 |
XP_016882772.1
|
5, 6 |
643 |
|
GenBank mRNA |
AK027901.1 |
- |
4, 5 |
511 |
|
AK096843.1 |
- |
3, 4 |
540 |
|
AK225793.1 |
- |
4, 5 |
483 |
|
AK291465.1 |
- |
4, 5 |
503 |
|
AK294543.1 |
- |
5, 6 |
587 |
|
BC001185.1 |
- |
4, 5 |
502 |
3502748 |
BC009335.2 |
- |
4, 5 |
502 |
4128019 |
CN301512.1 |
- |
4, 5 |
525 |
|
View Full Product Details