Gene Transcripts

Gene Symbol
Slc30a5
Entrez Gene ID
69048
Gene Name
solute carrier family 30 (zinc transporter), member 5
Gene Aliases
1810010K08Rik, AF233321, ZNT5, ZTL1, ZnT-5, Zntl1
Chromosome Location
Chr. 13: 100802645 - 100833563 on Build GRCm38
UniGene ID
Mm.402215
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022885.2 NP_075023.2 10 1357
GenBank mRNA AF233321.1 - 3 376
AF461761.1 - 10 1256
AK051183.1 - 10 1357
AK088004.1 - 9 1101
AK090176.1 - 10 1266
AK166006.1 - 10 1167
AK180687.1 - 2 88
BC029033.1 - 7 799
BC033452.1 - 10 1256 3156622

View Full Product Details