Gene Transcripts

Gene Symbol
Mrpl54
Entrez Gene ID
66047
Gene Name
mitochondrial ribosomal protein L54
Gene Aliases
0610008M19Rik, D10Sut1e
Chromosome Location
Chr. 10: 81264722 - 81266926 on Build GRCm38
UniGene ID
Mm.43397
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025317.2 NP_079593.1 1 129
GenBank mRNA AK002348.1 - 1 131
AK003225.1 - 1 130
AK009657.1 - 1 112
BC056985.1 - 1 112 6818054

View Full Product Details