Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025891.3 | NP_080167.3 | 11 | 1369 | |
GenBank mRNA | AK005094.1 | - | 11 | 1316 | |
AK019374.1 | - | 4 | 450 | ||
AK158019.1 | - | 1398 | |||
BC013122.1 | - | 11 | 1406 | 3586276 | |
BC060525.1 | - | 11 | 1268 | 6817219 |