Gene Transcripts
- Gene Symbol
- Enoph1
- Entrez Gene ID
- 67870
- Gene Name
- enolase-phosphatase 1
- Gene Aliases
- 2310057D15Rik, BB183658, C81437
- Chromosome Location
- Chr. 5: 100039994 - 100068925 on Build GRCm38
- UniGene ID
- Mm.11311
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163035.1 |
NP_001156507.1
|
1, 2 |
317 |
|
NM_026421.3 |
NP_080697.2
|
1, 2 |
317 |
|
GenBank mRNA |
AK009954.1 |
- |
1, 2 |
300 |
|
AK079062.1 |
- |
1, 2 |
311 |
|
AK080714.1 |
- |
1, 2 |
268 |
|
AK082747.1 |
- |
1, 2 |
292 |
|
AK152021.1 |
- |
1, 2 |
292 |
|
AK159356.1 |
- |
1, 2 |
290 |
|
AK167575.1 |
- |
1, 2 |
328 |
|
AK168696.1 |
- |
1, 2 |
279 |
|
AK177338.1 |
- |
1, 2 |
72 |
|
AK193770.1 |
- |
1, 2 |
72 |
|
BC021429.1 |
- |
1, 2 |
244 |
5030617 |
CX235072.1 |
- |
1, 2 |
235 |
|
View Full Product Details