Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026473.2 | NP_080749.2 | 4 | 1340 | |
GenBank mRNA | AK009958.1 | - | 4 | 1348 | |
AK015255.1 | - | 1 | 606 | ||
AK144815.1 | - | 4 | 1355 | ||
AK151643.1 | - | 4 | 1337 | ||
BC008225.1 | - | 4 | 1340 | 2631779 |