Gene Transcripts

Gene Symbol
Ngfr
Entrez Gene ID
18053
Gene Name
nerve growth factor receptor (TNFR superfamily, member 16)
Gene Aliases
LNGFR, Tnfrsf16, p75, p75NGFR, p75NTR
Chromosome Location
Chr. 11: 95568818 - 95587698 on Build GRCm38
UniGene ID
Mm.283893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033217.3 NP_150086.2 3 610
GenBank mRNA AF105292.1 - 3 468
AK037248.1 - 3 611
BC038365.1 - 3 601 5367638

View Full Product Details