Gene Transcripts
- Gene Symbol
- Nme7
- Entrez Gene ID
- 171567
- Gene Name
- NME/NM23 family member 7
- Gene Aliases
- D530024H21Rik, Nm23-M7, Nm23-r7
- Chromosome Location
- Chr. 1: 164307662 - 164437300 on Build GRCm38
- UniGene ID
- Mm.219428
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001293313.1 |
NP_001280242.1
|
5, 6 |
631 |
|
NM_138314.4 |
NP_612187.2
|
5, 6 |
634 |
|
NM_178071.6 |
NP_835172.1
|
5, 6 |
631 |
|
GenBank mRNA |
AF202048.1 |
- |
4, 5 |
535 |
|
AK044962.1 |
- |
5, 6 |
605 |
|
AK085222.1 |
- |
5, 6 |
579 |
|
AK133221.1 |
- |
|
1102 |
|
AK144915.1 |
- |
5, 6 |
635 |
|
AK164652.1 |
- |
5, 6 |
579 |
|
BC038021.1 |
- |
6, 7 |
661 |
3963844 |
BC049225.1 |
- |
4, 5 |
562 |
3483725 |
View Full Product Details