Gene Transcripts

Gene Symbol
Mettl13
Entrez Gene ID
71449
Gene Name
methyltransferase like 13
Gene Aliases
5630401D24Rik
Chromosome Location
Chr. 1: 162533671 - 162548345 on Build GRCm38
UniGene ID
Mm.270569
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144877.1 NP_659126.1 1 170
GenBank mRNA AK017470.1 - 1 355
AK048636.1 - 1 353
AK050492.1 - 1 363
AK084750.1 - 1 348
AK146751.1 - 368
AK162843.1 - 1 355
AK168959.1 - 1 377
AK169319.1 - 369
AK173044.1 - 323
BC014872.1 - 1 170 3602469

View Full Product Details