Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145384.2 | NP_663359.2 | 7 | 826 | |
GenBank mRNA | AK081661.1 | - | 7 | 813 | |
AK156668.1 | - | 7 | 826 | ||
BC019216.1 | - | 7 | 797 | 5065163 | |
BC025609.1 | - | 6 | 764 | 5309871 |