Gene Transcripts

Gene Symbol
Rasgef1b
Entrez Gene ID
320292
Gene Name
RasGEF domain family, member 1B
Gene Aliases
4732452O09Rik, GPIgamma4, Gpig4
Chromosome Location
Chr. 5: 99217420 - 99252927 on Build GRCm38
UniGene ID
Mm.259074
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145839.2 NP_665838.1 13 1582
NM_181318.4 NP_851835.1 13 1579
GenBank mRNA AK028756.1 - 13 1471
AK090085.1 - 13 1475
AK149812.1 - 13 1501
AY129963.1 - 13 1509
BC050858.1 - 13 1512 6517502
BC052726.1 - 13 1473 6813357

View Full Product Details