Gene Transcripts

Gene Symbol
Kcnj14
Entrez Gene ID
211480
Gene Name
potassium inwardly-rectifying channel, subfamily J, member 14
Gene Aliases
A930026G01Rik, IRK4, Kir2.4
Chromosome Location
Chr. 7: 45816467 - 45824747 on Build GRCm38
UniGene ID
Mm.68170
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145963.2 NP_666075.1 3 2177
GenBank mRNA AK030711.1 - 3 2187
AK044725.1 - 3 2158
AK080732.1 - 3 2166
BC022700.1 - 1 1276
BC029692.1 - 3 2117 5363132
BC031753.1 - 3 2177 4503027

View Full Product Details