Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000979.3 | NP_000970.1 | 2, 3 | 360 | |
GenBank mRNA | AK026558.1 | - | 2, 3 | 118 | |
AK293386.1 | - | 2, 3 | 119 | ||
BC000374.2 | - | 1, 2 | 92 | 2820119 | |
BC009708.2 | - | 2, 3 | 106 | 3878799 | |
BC021743.2 | - | 2, 3 | 118 | 5261251 | |
BM764512.1 | - | 2, 3 | 478 | ||
DQ403032.1 | - | 1, 2 | 16 | ||
L11566.1 | - | 2, 3 | 100 |