Gene Transcripts
- Gene Symbol
- Nrbp1
- Entrez Gene ID
- 192292
- Gene Name
- nuclear receptor binding protein 1
- Gene Aliases
- B230344L17Rik, Nrbp
- Chromosome Location
- Chr. 5: 31228089 - 31251566 on Build GRCm38
- UniGene ID
- Mm.292040
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_147201.2 |
NP_671734.1
|
2, 3 |
322 |
|
GenBank mRNA |
AF302138.1 |
- |
2, 3 |
321 |
|
AK046142.1 |
- |
2, 3 |
316 |
|
AK077440.1 |
- |
2, 3 |
358 |
|
AK170501.1 |
- |
2, 3 |
338 |
|
AK178908.1 |
- |
2, 3 |
163 |
|
AK191367.1 |
- |
3, 4 |
202 |
|
AK217229.1 |
- |
1, 2 |
249 |
|
BC004756.1 |
- |
2, 3 |
292 |
3154089 |
BC018463.1 |
- |
2, 3 |
296 |
3598438 |
View Full Product Details