Gene Transcripts

Gene Symbol
RPS25
Entrez Gene ID
6230
Gene Name
ribosomal protein S25
Gene Aliases
S25
Chromosome Location
Chr. 11: 119015712 - 119018347 on Build GRCh38
UniGene ID
Hs.512676
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001028.2 NP_001019.1 3 251
GenBank mRNA AK311883.1 - 3 251
BC004294.1 - 1 279 3622031
BC004986.1 - 3 234 2905996
M64716.1 - 3 259

View Full Product Details