Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001072.3 | NP_001063.2 | 1 | 973 | |
NM_205862.1 | NP_995584.1 | 2 | 186 | ||
GenBank mRNA | AY435141.1 | - | 1 | 834 | |
BC019861.1 | - | 1 | 868 | 4454322 | |
BM688740.1 | - | 2 | 186 | ||
BM924331.1 | - | 2 | 204 | ||
DQ364250.1 | - | 1 | 842 | ||
J04093.1 | - | 1 | 918 |