Gene Transcripts

Gene Symbol
Ttll8
Entrez Gene ID
239591
Gene Name
tubulin tyrosine ligase-like family, member 8
Gene Aliases
1700019P01Rik, 4933401B17
Chromosome Location
Chr. 15: 88889778 - 88955641 on Build GRCm38
UniGene ID
Mm.479911
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172818.3 NP_766406.2 5 863
GenBank mRNA AK030151.1 - 5 856
AM690752.1 - 5 520
BC116293.1 - 5 504 40094952

View Full Product Details