Gene Transcripts

Gene Symbol
Hpcal4
Entrez Gene ID
170638
Gene Name
hippocalcin-like 4
Gene Aliases
AI846570, NVP-2
Chromosome Location
Chr. 4: 123183504 - 123194699 on Build GRCm38
UniGene ID
Mm.270641
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174998.4 NP_778163.1 5 1171
GenBank mRNA AK032591.1 - 5 1170
BC138631.1 - 4 1006 8861651
BC138632.1 - 4 1006 8861652

View Full Product Details