Gene Transcripts

Gene Symbol
TNC
Entrez Gene ID
3371
Gene Name
tenascin C
Gene Aliases
150-225, DFNA56, GMEM, GP, HXB, JI, TN, TN-C
Chromosome Location
Chr. 9: 115019575 - 115118257 on Build GRCh38
UniGene ID
Hs.143250
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002160.3 NP_002151.2 17 5487
XM_005251972.3 XP_005252029.1 16 5164
XM_005251974.3 XP_005252031.1 11 3799
XM_006717096.3 XP_006717159.1 18 5713
XM_006717097.3 XP_006717160.1 16 5164
XM_006717098.3 XP_006717161.1 15 4891
XM_011518626.2 XP_011516928.1 14 4621
XM_011518629.2 XP_011516931.1 12 4072
XM_017014678.1 XP_016870167.1 19 5986
XM_017014679.1 XP_016870168.1 18 5713
XM_017014680.1 XP_016870169.1 18 5710
XM_017014681.1 XP_016870170.1 15 4894
GenBank mRNA AB210038.1 - 17 5363
GU473235.1 - 12 3986
M24630.1 - 15 3785
M55618.1 - 16 5210
X56160.1 - 16 5126
X78565.1 - 17 5388

View Full Product Details