Gene Transcripts

Gene Symbol
LIG3
Entrez Gene ID
3980
Gene Name
DNA ligase 3
Gene Aliases
LIG2
Chromosome Location
Chr. 17: 34980498 - 35009169 on Build GRCh38
UniGene ID
Hs.100299
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002311.4 NP_002302.2 7 1355
NM_013975.3 NP_039269.2 7 1355
XM_005257970.3 XP_005258027.1 7 1323
XM_006721896.3 XP_006721959.1 7 1324
XM_017024624.1 XP_016880113.1 7 1519
GenBank mRNA AK300363.1 - 7 1315
AK308407.1 - 7 1300
BC068005.1 - 7 1330 6092747
U40671.1 - 7 1316
X84740.1 - 7 1298

View Full Product Details