Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000962.3 | NP_000953.2 | 11 | 1573 | |
NM_001271164.1 | NP_001258093.1 | 10 | 1429 | ||
NM_001271165.1 | NP_001258094.1 | 10 | 1522 | ||
NM_001271166.1 | NP_001258095.1 | 11 | 1688 | ||
NM_001271367.1 | NP_001258296.1 | 11 | 1433 | ||
NM_001271368.1 | NP_001258297.1 | 12 | 1910 | ||
NM_080591.2 | NP_542158.1 | 11 | 1462 | ||
XM_005252105.3 | XP_005252162.1 | 12 | 1748 | ||
XM_011518875.2 | XP_011517177.1 | 12 | 2009 | ||
XM_011518876.2 | XP_011517178.1 | 12 | 5619 | ||
XM_017014929.1 | XP_016870418.1 | 14 | 1983 | ||
GenBank mRNA | AK290022.1 | - | 11 | 1526 | |
AK295221.1 | - | 12 | 1895 | ||
AK304403.1 | - | 10 | 1522 | ||
AK308222.1 | - | 11 | 1687 | ||
BC029840.1 | - | 11 | 1545 | 5174923 | |
CD014048.1 | - | 10 | 1395 | ||
CD014049.1 | - | 11 | 1399 | ||
DQ180741.1 | - | 10 | 1586 | ||
DQ180742.1 | - | 10 | 1586 | ||
M59979.1 | - | 10 | 1498 | ||
S36219.1 | - | 10 | 1382 | ||
S36271.1 | - | 10 | 1493 | ||
S78220.1 | - | 10 | 1493 |