Gene Transcripts

Gene Symbol
PWP2
Entrez Gene ID
5822
Gene Name
PWP2 periodic tryptophan protein homolog (yeast)
Gene Aliases
EHOC-17, PWP2H, UTP1
Chromosome Location
Chr. 21: 44107262 - 44131181 on Build GRCh38
UniGene ID
Hs.449076
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005049.2 NP_005040.2 13 1669
XM_011529667.2 XP_011527969.1 13 1734
GenBank mRNA BM457858.1 - 4 502

View Full Product Details