Gene Transcripts

Gene Symbol
SLC11A1
Entrez Gene ID
6556
Gene Name
solute carrier family 11 member 1
Gene Aliases
LSH, NRAMP, NRAMP1
Chromosome Location
Chr. 2: 218381766 - 218396894 on Build GRCh38
UniGene ID
Hs.591607
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000578.3 NP_000569.3 9 1222
XM_005246793.3 XP_005246850.1 8 1342
XM_005246794.3 XP_005246851.1 10 1559
XM_006712709.3 XP_006712772.1 9 1416
XM_006712710.3 XP_006712773.1 8 1293
XM_006712711.3 XP_006712774.1 7 1219
XM_011511684.2 XP_011509986.1 10 1594
XM_011511685.2 XP_011509987.1 9 1451
XM_017004765.1 XP_016860254.1 8 1362
XM_017004766.1 XP_016860255.1 9 1453
GenBank mRNA AB209650.1 - 7 2936
AK298667.1 - 9 976
AK303398.1 - 9 946
AK307954.1 - 9 1162
AK310914.1 - 9 978
AK311405.1 - 8 829
BC033754.1 - 7 1228
BC071165.1 - 10 1016 5756382
D38171.1 - 7 729
D50402.1 - 9 1057
D50403.1 - 9 968
L32185.1 - 9 958

View Full Product Details