Gene Transcripts
- Gene Symbol
- ITGA10
- Entrez Gene ID
- 8515
- Gene Name
- integrin subunit alpha 10
- Gene Aliases
- PRO827
- Chromosome Location
- Chr. 1: 145891208 - 145910189 on Build GRCh38
- UniGene ID
- Hs.158237
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001303040.1 |
NP_001289969.1
|
23, 24 |
2970 |
|
NM_001303041.1 |
NP_001289970.1
|
22, 23 |
2858 |
|
NM_003637.4 |
NP_003628.2
|
26, 27 |
3287 |
|
XM_005277436.3 |
XP_005277493.1
|
25, 26 |
3038 |
|
XM_011510083.2 |
XP_011508385.1
|
22, 23 |
2718 |
|
XM_011510084.1 |
XP_011508386.1
|
15, 16 |
1715 |
|
XM_017002622.1 |
XP_016858111.1
|
24, 25 |
3244 |
|
XM_017002623.1 |
XP_016858112.1
|
24, 25 |
3241 |
|
XM_017002625.1 |
XP_016858114.1
|
23, 24 |
3130 |
|
XM_017002626.1 |
XP_016858115.1
|
22, 23 |
3001 |
|
XM_017002628.1 |
XP_016858117.1
|
20, 21 |
2694 |
|
GenBank mRNA |
AF074015.1 |
- |
26, 27 |
3133 |
|
AF112345.1 |
- |
26, 27 |
3188 |
|
AK304155.1 |
- |
23, 24 |
2826 |
|
AK314255.1 |
- |
26, 27 |
3136 |
|
BC140831.1 |
- |
22, 23 |
2733 |
9021699 |
BC144637.1 |
- |
22, 23 |
2733 |
9053168 |
View Full Product Details