Gene Transcripts

Gene Symbol
LHFPL2
Entrez Gene ID
10184
Gene Name
lipoma HMGIC fusion partner-like 2
Gene Aliases
Chromosome Location
Chr. 5: 78485215 - 78648895 on Build GRCh38
UniGene ID
Hs.670094
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005779.2 NP_005770.1 5 1123
XM_006714515.1 XP_006714578.1 4 810
GenBank mRNA AK125227.1 - 2 620
AY309920.1 - 2 522
BC136335.1 - 2 474 9020322
BC136336.1 - 2 474 9020323
CR936660.1 - 3 760
D86961.1 - 2 342

View Full Product Details