Gene Transcripts

Gene Symbol
UGT2A1
Entrez Gene ID
10941
Gene Name
UDP glucuronosyltransferase family 2 member A1 complex locus
Gene Aliases
UDPGT2A1
Chromosome Location
Chr. 4: 69588417 - 69653249 on Build GRCh38
UniGene ID
Hs.225950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252274.2 NP_001239203.2 2 780
NM_001252275.2 NP_001239204.2 2 780
NM_001301239.1 NP_001288168.1 1 718
NM_006798.4 NP_006789.3 2 780
GenBank mRNA AJ006054.1 - 1 727
AK304249.1 - 2 780
AK314209.1 - 2 778
BC142640.1 - 1 781

View Full Product Details