Gene Transcripts

Gene Symbol
SAMD4B
Entrez Gene ID
55095
Gene Name
sterile alpha motif domain containing 4B
Gene Aliases
SMGB, Smaug2
Chromosome Location
Chr. 19: 39342459 - 39390748 on Build GRCh38
UniGene ID
Hs.612332
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303614.1 NP_001290543.1 5, 6 1338
NM_018028.3 NP_060498.2 8, 9 2059
XM_011527057.2 XP_011525359.1 6, 7 1349
XM_011527060.2 XP_011525362.1 6, 7 2451
XM_011527061.2 XP_011525363.1 5, 6 1263
XM_011527062.2 XP_011525364.1 5, 6 1324
XM_011527063.2 XP_011525365.1 6, 7 1348
XM_011527064.2 XP_011525366.1 6, 7 1349
XM_017026918.1 XP_016882407.1 5, 6 1283
XM_017026919.1 XP_016882408.1 7, 8 1439
XM_017026920.1 XP_016882409.1 7, 8 1461
XM_017026921.1 XP_016882410.1 5, 6 1324
XM_017026922.1 XP_016882411.1 7, 8 1441
XM_017026923.1 XP_016882412.1 7, 8 6979
GenBank mRNA AK001073.1 - 224
BC039652.1 - 1, 2 36
BC054518.1 - 8, 9 2050 6501489
BC065211.1 - 2, 3 346
BC080641.1 - 5, 6 1298 6645022
EF601121.1 - 8, 9 2046

View Full Product Details