Gene Transcripts

Gene Symbol
VPS13D
Entrez Gene ID
55187
Gene Name
vacuolar protein sorting 13 homolog D
Gene Aliases
Chromosome Location
Chr. 1: 12230039 - 12512047 on Build GRCh38
UniGene ID
Hs.439381
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015378.3 NP_056193.2 29 7016
NM_018156.3 NP_060626.2 29 7016
GenBank mRNA AB007922.3 - 11 3326
AJ608774.1 - 29 6988
AJ608775.1 - 29 6988
AK001481.1 - 2 136
AK125118.1 - 5 746
BX641035.1 - 4 580

View Full Product Details