Gene Transcripts
- Gene Symbol
- LSM2
- Entrez Gene ID
- 57819
- Gene Name
- LSM2 homolog, U6 small nuclear RNA and mRNA degradation associated
- Gene Aliases
- C6orf28, G7B, YBL026W, snRNP
- Chromosome Location
- Chr. 6: 31797392 - 31806984 on Build GRCh38
- UniGene ID
- Hs.103106
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021177.4 |
NP_067000.1
|
3, 4 |
323 |
|
GenBank mRNA |
AF136977.1 |
- |
3, 4 |
308 |
|
AF182288.1 |
- |
3, 4 |
142 |
|
AF196468.1 |
- |
3, 4 |
288 |
|
AJ245416.1 |
- |
3, 4 |
314 |
|
BC009192.2 |
- |
3, 4 |
305 |
3842537 |
BP203102.1 |
- |
3, 4 |
323 |
|
BU189898.1 |
- |
3, 4 |
314 |
|
CR542146.1 |
- |
2, 3 |
96 |
|
CR542157.1 |
- |
2, 3 |
96 |
|
View Full Product Details