Gene Transcripts

Gene Symbol
NOX5
Entrez Gene ID
79400
Gene Name
NADPH oxidase, EF-hand calcium binding domain 5
Gene Aliases
Chromosome Location
Chr. 15: 68930500 - 69057161 on Build GRCh38
UniGene ID
Hs.657932
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184779.1 NP_001171708.1 16 2156
NM_001184780.1 NP_001171709.1 16 2283
NM_024505.3 NP_078781.3 16 2240
NR_033671.2 17 2471
NR_033672.1 17 2387
GenBank mRNA AF317889.1 - 13 1671
AF325189.1 - 17 2362
AF325190.1 - 17 2446
AF353088.1 - 16 2135
AF353089.1 - 16 2219
AK026011.1 - 13 1725
AK074058.1 - 14 1909
AK074071.1 - 15 5358
AK314689.1 - 16 2219
AY166601.1 - 12 1466
BC125097.1 - 16 2149 40118286
BC125098.1 - 17 2253 40118288
DQ413001.1 - 13 1599

View Full Product Details