Gene Transcripts
- Gene Symbol
- TUBA4B
- Entrez Gene ID
- 80086
- Gene Name
- tubulin alpha 4b
- Gene Aliases
- TUBA4
- Chromosome Location
- Chr. 2: 219253243 - 219272188 on Build GRCh38
- UniGene ID
- Hs.664469
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003063.1 |
|
2, 3 |
228 |
|
GenBank mRNA |
AK024002.1 |
- |
2, 3 |
228 |
|
View Full Product Details