Gene Transcripts

Gene Symbol
GPAT3
Entrez Gene ID
84803
Gene Name
glycerol-3-phosphate acyltransferase 3
Gene Aliases
AGPAT 10, AGPAT10, AGPAT8, AGPAT9, HMFN0839, LPAAT-theta, MAG1
Chromosome Location
Chr. 4: 83535634 - 83605875 on Build GRCh38
UniGene ID
Hs.99196
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256421.1 NP_001243350.1 9, 10 1136
NM_001256422.1 NP_001243351.1 9, 10 1140
NM_032717.4 NP_116106.2 8, 9 1223
XM_011532383.1 XP_011530685.1 7, 8 1029
XM_011532384.2 XP_011530686.1 9, 10 3224
XM_017008780.1 XP_016864269.1 9, 10 1292
XM_017008781.1 XP_016864270.1 9, 10 1299
GenBank mRNA AB075872.1 - 5, 6 421
AK055749.1 - 9, 10 1135
AK223543.1 - 9, 10 1107
AY358100.1 - 8, 9 1040
BC073136.1 - 3, 4 199
BC090956.1 - 8, 9 1129 30387839
DQ324782.1 - 8, 9 920
DQ345298.1 - 8, 9 1220

View Full Product Details