Gene Transcripts
- Gene Symbol
- POC1B
- Entrez Gene ID
- 282809
- Gene Name
- POC1 centriolar protein B
- Gene Aliases
- CORD20, PIX1, TUWD12, WDR51B
- Chromosome Location
- Chr. 12: 89419718 - 89526262 on Build GRCh38
- UniGene ID
- Hs.25130
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199777.1 |
NP_001186706.1
|
6, 7 |
1168 |
|
NM_172240.2 |
NP_758440.1
|
7, 8 |
1187 |
|
NR_037659.1 |
|
6, 7 |
1015 |
|
NR_037660.1 |
|
5, 6 |
996 |
|
GenBank mRNA |
AB053266.1 |
- |
6, 7 |
677 |
|
AK027829.1 |
- |
3, 4 |
386 |
|
AK074772.1 |
- |
7, 8 |
925 |
|
AK123625.1 |
- |
3, 4 |
385 |
|
AK295596.1 |
- |
6, 7 |
753 |
|
AL832918.1 |
- |
5, 6 |
652 |
|
BC026080.2 |
- |
7, 8 |
949 |
4827689 |
BU521431.1 |
- |
5, 6 |
665 |
|
EF011619.1 |
- |
7, 8 |
949 |
|
View Full Product Details