Gene Transcripts

Gene Symbol
OR6S1
Entrez Gene ID
341799
Gene Name
olfactory receptor family 6 subfamily S member 1
Gene Aliases
OR14-37, OR6S1Q
Chromosome Location
Chr. 14: 20640696 - 20641691 on Build GRCh38
UniGene ID
Hs.513132
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001968.1 NP_001001968.1 1 877

View Full Product Details