Gene Transcripts

Gene Symbol
RING1
Entrez Gene ID
6015
Gene Name
ring finger protein 1
Gene Aliases
RING1A, RNF1
Chromosome Location
Chr. 6: 33208509 - 33212722 on Build GRCh38
UniGene ID
Hs.631989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002931.3 NP_002922.2 2 242
GenBank mRNA AK289355.1 - 2 251
AK297552.1 - 2 241
AK301010.1 - 2 251
AK301378.1 - 2 245
BC002922.2 - 2 237 3954313
BC051866.2 - 2 235 6142438
BI259576.1 - 2 251

View Full Product Details