Gene Transcripts

Gene Symbol
SMARCD3
Entrez Gene ID
6604
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3
Gene Aliases
BAF60C, CRACD3, Rsc6p
Chromosome Location
Chr. 7: 151238973 - 151277436 on Build GRCh38
UniGene ID
Hs.647067
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003801.1 NP_001003801.1 12 1478
NM_001003802.1 NP_001003802.1 13 1467
NM_003078.3 NP_003069.2 13 1506
XM_011516521.2 XP_011514823.1 12 1585
XM_017012556.1 XP_016868045.1 12 1321
GenBank mRNA AK127595.1 - 13 2014
AY450430.1 - 12 1782
AY450431.1 - 13 1467
BC002628.2 - 13 1467 3608512
BC110350.1 - 12 1397 5212363
BX648385.1 - 11 2229
U66619.1 - 13 1515

View Full Product Details