Gene Transcripts

Gene Symbol
EIF3J
Entrez Gene ID
8669
Gene Name
eukaryotic translation initiation factor 3 subunit J
Gene Aliases
EIF3S1, eIF3-alpha, eIF3-p35
Chromosome Location
Chr. 15: 44537068 - 44562803 on Build GRCh38
UniGene ID
Hs.404056
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284335.1 NP_001271264.1 5 474
NM_001284336.1 NP_001271265.1 3 327
NM_003758.3 NP_003749.2 5 474
GenBank mRNA AF090923.1 - 1 70
AK002006.1 - 5 394
AK300659.1 - 5 359
AL536561.3 - 3 204
BC002719.2 - 5 407 3632801
U97670.1 - 5 407

View Full Product Details