Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021067.4 | NP_066545.3 | 6 | 610 | |
NR_134574.1 | 7 | 732 | |||
XM_017028162.1 | XP_016883651.1 | 6 | 567 | ||
GenBank mRNA | AK294593.1 | - | 7 | 686 | |
BC012542.1 | - | 6 | 611 | 4333095 | |
D80008.1 | - | 6 | 566 |